Key Documents
57548
Low Volume Static Mixer (50-250 μL)
housing, In-line, stainless steel
Select a Size
1 EA
$739.00
$739.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$739.00
About This Item
UNSPSC Code:
41115854
$739.00
Please contact Customer Service for Availability
Request a Bulk Order
material
stainless steel
compatibility
for use with ASI low volume mixer cartridge
General description
Low Volume Mixer Housing (housing, In-Line) is an accessory for use with low volume mixer cartridge (50-250 μL) in ASI Static Mixers, used for HPLC and post-column derivatization. “Inline” refers to a single line going into the mixer housing. It is mainly used when additional mixing is needed in a single flow-path.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)