Key Documents
57513-U
Millipore
Porozorb Cartridge
Porozorb 4004, size 4000 mL
Select a Size
1 EA
$5,871.00
$5,871.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$5,871.00
About This Item
UNSPSC Code:
23151817
NACRES:
SB.52
$5,871.00
Please contact Customer Service for Availability
Quality Level
size
4000 mL
General description
The Porozorb cartridge is a ready-to-use column containing Amberlite XAD®-4 polymeric hydrophobic resin. It removes detergents effectively, as well as isolates and/or purifies pharmaceutical and biological compounds. This cartridge is suitable for removing phenols and other organics in aqueous solutions, and for recovering antibodies.
Other Notes
Not for clinical or diagnostic use. Due to shelf life limitations, the cartridges are made upon receipt of an order; expect a 2-4 week delay in shipping times. Determination of cartridge purity for a particular application is the purchaser′s responsibility. Equilibrate the cartridge with matrix buffer prior to use.
Legal Information
Amberlite is a trademark of DuPont de Nemours, Inc.
Porozorb is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Choose from one of the most recent versions:
Certificates of Analysis (COA)