Key Documents
57418
Supelco
Gripper fitting
for use with 1/8 in. tubing
Select a Size
10 EA
$95.10
$95.10
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
10 EA
$95.10
About This Item
UNSPSC Code:
12000000
$95.10
Please contact Customer Service for Availability
Request a Bulk Order
description
Tube end fittings (nuts) not included.
packaging
pkg of 10 ea
technique(s)
LPLC: suitable
compatibility
for use with 1/8 in. tubing
for use with Omnifit fittings
General description
Gripper fitting is used in sealing of the protein chip cartridge by screwing the plastic holder and connection tubing.[1]
Application
Gripper fitting may be used in chromatographic sample preparation, purification and traditional/classical flash chromatography.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Kevin A Heyries et al.
Biosensors & bioelectronics, 23(12), 1812-1818 (2008-04-09)
Protein microarrays for allergen-specific antibodies detection were integrated in microfluidic chips, with imaging chemiluminescence as the analytical technique. This paper demonstrates the feasibility of miniaturized chemiluminescent ELISA by presenting rapid, reproducible and sensitive detection of protein antibodies using microfluidics. Three