Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57273-U

Supelco

Visiprep Large Volume Sampler Tube Adapters, 1/4-28 Threads

for use with 3 or 6 mL SPE tubes, pk of 4

Synonym(s):

PK4 3/6ML TUBE ADAPTER W/THREADS FOR LARGE VOLUME SAMPLER

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
$103.00

$103.00


Available to ship onApril 23, 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
4 EA
$103.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

$103.00


Available to ship onApril 23, 2025Details


Request a Bulk OrderRequest a Sample

packaging

pk of 4

technique(s)

solid phase extraction (SPE): suitable

fitting

male adapter for 3 or 6 mL SPE tube
1/4-28 female UNF bore

application(s)

food and beverages

compatibility

for use with 3 or 6 mL SPE tubes

Looking for similar products? Visit Product Comparison Guide

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service