Skip to Content
MilliporeSigma
All Photos(1)

Documents

57162-U

Supelco

Preppy Vacuum Manifold Collection Rack

for use with 15 mL (21 mm OD x 70 mm H) or 20 mL (28 mm OD x 61 mm H) vials.

Synonym(s):

Preppy 12 PORT RACK PLATE, 15 & 20ML VIALS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with 15 mL (21 mm OD x 70 mm H) or 20 mL (28 mm OD x 61 mm H) vials.

Looking for similar products? Visit Product Comparison Guide

Application

Recommended 15 mL vials compatible with this product: Cat. No. 27159, 27008, 27014-U. Recommended 20 mL vials compatible with this product: Cat. No. Z190527, 190523-1PAK.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service