Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57159-U

Supelco

Preppy Vacuum Manifold Collection Rack

for use with 2 mL (12 mm OD x 32 mm H) or 4 mL (15 mm OD x 45 mm H) vials

Synonym(s):

Preppy 12 PORT RACK PLATE, 1 & 4ML VIALS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

technique(s)

solid phase extraction (SPE): suitable

Quality Level

application(s)

food and beverages

compatibility

for use with 2 mL (12 mm OD x 32 mm H) or 4 mL (15 mm OD x 45 mm H) vials

Looking for similar products? Visit Product Comparison Guide

Application

Recommended 2 mL vials compatible with this product: Cat. No. 27487-U, 27490-U, or 27493. Recommended 4 mL vials compatible with this product: Cat. No. 27136, 27006, 27012.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service