Skip to Content
MilliporeSigma
All Photos(1)

Documents

55519-U

Supelco

SupelGenie RP-Amide Online SPE Cartridges

L × I.D. 20 mm × 4.0 mm, pkg of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.21

material

stainless steel tube

product line

Supel

composition

bed wt., 180 mg

packaging

pkg of 2 ea

technique(s)

solid phase extraction (SPE): suitable

L × I.D.

20 mm × 4.0 mm

matrix active group

RP-Amide

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices. This technique is typically performed manually (offline). Offline SPE is generally time-consuming, labor intensive, irreproducible from batch to batch, and incompatible with automation.

Supel Genie Online SPE cartridges are designed for direct injection of untreated samples (e.g. water and urine) or processed samples (e.g. protein-precipitated plasma and serum), for analysis with liquid chromatography and mass spectrometry (LC/MS). The Supel Genie Online SPE cartridges enable online sample cleanup, concentration, and separation with excellent reproducibility and performance. Coupled with mass spectrometry for detection, the Supel Genie Online SPE cartridges provide a simple, automated method for sample analysis and are ideal for use in the LC/MS/MS analysis of biological fluids such as plasma and serum.

For first time use, a starter kit is required to obtain reusable universal hardware that will fit all Supel Genie Online SPE cartridges (P/N 55516-U).

LC instrumentation must be configured to accommodate Online SPE cartridges. Please see instrument setup diagram in datasheet. 55519-U

Please contact techserv@sial.com for more information on setup and hardware.

Application

Sample Preparation, HPLC

Features and Benefits

  • Provides cleaner samples for longer column life, less instrument downtime, and more accurate and reproducible data
  • Automation saves time, decreases costs, and eliminates human error
  • Highly reproducible, consistent product performance
  • RP-Amide cartridges often offer improved performance for polar analytes, especially those that can interact via hydrogen bonding, when compared to standard alkyl phases
  • Cartridges also available in pack of 6

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service