Key Documents
55051
RheBuild® Kit
for use with 9125/9126
Select a Size
1 EA
$341.00
$341.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$341.00
About This Item
UNSPSC Code:
41115854
$341.00
Please contact Customer Service for Availability
Request a Bulk Order
compatibility
for use with 9125/9126
General description
These kits include all of the tools and parts you need to repair your Rheodyne injector. The parts included are those most likely to be damaged, worn, or lost. Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
RheBuild is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)