Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54922

Supelco

Nucleosil® C8 HPLC Column

5 μm particle size, L × I.D. 25 cm × 3.2 mm

Synonym(s):

Nucleosil® C8, 5UM, 25CM X 3.2MM ID

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Agency

suitable for USP L7

feature

endcapped: no

manufacturer/tradename

Nucleosil®

extent of labeling

8.5% Carbon loading

parameter

400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 3.2 mm

surface area

350 m2/g

matrix

octyl

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

100 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service