Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54809

Sigma-Aldrich

Mono P® 5/50 GL

10 μm particle size, L × I.D. 5 cm × 5 mm

Synonym(s):

Amersham Biosciences Ion Exchange Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

L × I.D.

5 cm × 5 mm

matrix

MonoBeads

matrix active group

amphoteric

particle size

10 μm

compatibility

mode of use ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Mono columns are highly efficient, pH-stable columns designed for high performance ion exchange separations of proteins, peptides, and polynucleotides, in applications including peptide mapping and monoclonal antibody purification. The unique properties of these columns are based on MonoBeads support – a beaded hydrophilic material with the narrowest particle size distribution of any chromatographic support. This monodispersity permits high flow rates at relatively low backpressures.

Application

Mono columns are used in peptide mapping and monoclonal antibody purification. Mono P columns have been used to purify homogenous forms of fungal peroxygenase and to investigate separation of DNA restriction fragments.

Legal Information

Mono P is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

René Ullrich et al.
Biotechnology journal, 4(11), 1619-1626 (2009-07-09)
Extracellular peroxygenase from the agaric fungus Agrocybe aegerita is a versatile biocatalyst that oxygenates various substrates by means of hydrogen peroxide. The enzyme is routinely produced in suspensions of soybean meal and has until now been purified by several steps
E Westman et al.
Analytical biochemistry, 166(1), 158-171 (1987-10-01)
Separation of DNA restriction fragments by FPLC ion-exchange chromatography on Mono Q and Mono P columns was investigated. The columns were found to be particularly suitable for the separation of fragments up to 500-600 bp long. Larger fragments can also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service