Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54277-U

Supelco

SupelMIP® SPE - Bisphenol A

bed weight 100 mg, volume 6 mL, pk of 50

Synonym(s):

SupelMIP® SPE - Bisphenol A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

polypropylene tube

composition

bed weight, 100 mg

packaging

pk of 50

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

matrix active group

bisphenol A MIP coating

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.

By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection.

Visit www.sigma-aldrich.com/supelmip to learn more.

Application

Developed for the class-selective extraction of Bisphenol A (BPA) from broth or milk-based matrices.

Features and Benefits

  • Highly selective SPE for trace analysis in complex matrices
  • Achieve lower detection limits
  • Significant time and cost savings
  • Improved MS-compatibility

Legal Information

SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

440.6 °F - closed cup

Flash Point(C)

227 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

SupelMIP SPE is based on molecularly imprinted polymer (MIP) technology. Each SupelMIP phase offers tailor-made selectivity for the extraction of trace analytes in complex matrixes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service