Skip to Content
MilliporeSigma
All Photos(1)

Documents

54058-U

Supelco

Supelclean ENVI-Carb II/PSA SPE Tube

bed B 600 mg (Supelclean PSA SPE), bed A 300 mg (Supelclean ENVI-Carb-II SPE), volume 6 mL, pk of 30

Synonym(s):

Carbon/PSA SPE cartridge tube, 6 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

PE frit (20 μm porosity)
polypropylene hardware

product line

Supelclean

composition

bed A, 300 mg (Supelclean ENVI-Carb-II SPE)
bed B, 600 mg (Supelclean PSA SPE)

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

matrix active group

PSA phase
carbon phase

application(s)

food and beverages

separation technique

ion exchange
reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Reversed-phase and anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • Dual layer SPE tube that contains both Supelclean ENVI-Carb (upper layer) & PSA (lower layer) SPE sorbents (separated by PE frit)
  • Developed to offer superior clean up when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.).
  • ENVI-carb has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products
  • Supelclean PSA is a polymerically bonded, ethylenediamine-N-propyl phase that contains both primary and secondary amines
  • Supelclean PSA has a strong affinity and high capacity for fatty acids, organic acids, and some polar pigments and sugars
  • Tested for superior cleanliness using GC-FID and GC-MS

Application


  • Pipette-tip SPE based on Graphene/ZnCr LDH for Pb(II) analysis in hair samples followed by GFAAS.: This research highlights the application of Supelclean ENVI-Carb II/PSA SPE Tubes in the extraction and analysis of lead(II) ions from hair samples. The tubes significantly improve the efficiency of the sample preparation process, enabling precise and reliable lead quantification using graphite furnace atomic absorption spectroscopy (GFAAS) (Mirzaee et al., 2021).

Legal Information

ENVI-Carb is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversed-phase interaction will retain most molecules with hydrophobic character; it is very useful for extracting analytes that are very diverse in structure within the same sample.

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service