Skip to Content
MilliporeSigma
All Photos(3)

Documents

53225-U

Supelco

SupelMIP® SPE - Beta-agonists

bed wt. 25 mg, volume 3 mL, pk of 50

Synonym(s):

MIP SPE (molecular imprinted) Beta-agonist cartridge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

polypropylene tube

product line

SupelMIP®

composition

bed wt., 25 mg

packaging

pk of 50

technique(s)

solid phase extraction (SPE): suitable

volume

3 mL

matrix active group

SupelMIP® phase

Looking for similar products? Visit Product Comparison Guide

General description

SupelMIP SPE phases were developed by MIP Technologies AB, which is one of the leading authorities and commercial pioneers of molecular imprinted polymers for process scale separations, analytical chromatography, and sample preparation.

The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.

By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection.

Visit www.sigma-aldrich.com/supelmip to learn more.

Application

A class selective MIP phase for the multi-residue extraction of beta-agonists in urine, tissues, and other biological fluids.

SupelMIP - Beta-agonists was developed for the selective extaction of a broad range o beta-agonists such as brombuterol, clenbuterol, formoterol, isoxuprine, mapenterol, ractopamine, ritrodine, salbutamol, salmeterol, terbutaline, tulobuterol, zilpaterol and others. The SupelMIP cartridge offers excellent reproducibility for surveillance of this common class of drugs of abuse. Loading capacity is superior to immuno-affinity columns, and MS-detection is recommended for forensic validity. Detailed extraction protocols are included with the product.

Features and Benefits

  • Highly selective SPE for trace analysis in complex matrices
  • Achieve lower detection limits
  • Significant time and cost savings
  • Improved MS-compatibility

Legal Information

SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

200.3 °F

Flash Point(C)

93.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

SupelMIP SPE is based on molecularly imprinted polymer (MIP) technology. Each SupelMIP phase offers tailor-made selectivity for the extraction of trace analytes in complex matrixes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service