Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506877

Supelco

2-Chlorobenzoic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57
Beilstein:
907340
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

138-140 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

OC(=O)c1ccccc1Cl

InChI

1S/C7H5ClO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)

InChI key

IKCLCGXPQILATA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chlorobenzoic acid may be used as a reference standard in the determination of 2-chlorobenzoic acid in water samples using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

343.4 °F

Flash Point(C)

173 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental design for the study of two derivatization procedures for simultaneous GC analysis of acidic herbicides and water chlorination by-products.
Boucharat C, et al.
Talanta, 47(2), 311-323 (1998)
B R Genthner
Biodegradation, 10(1), 27-33 (1999-07-29)
Dechlorination was the initial step of 2CB biodegradation in four 2-chlorobenzoate-degrading methanogenic consortia. Selected characteristics of ortho reductive dehalogenation were examined in consortia developed from the highest actively dechlorinating dilutions of the original 2CB consortia, designated consortia M34(-9), P20(-9), P21(-9)
Gang-hua Lang et al.
Applied microbiology and biotechnology, 83(6), 1085-1094 (2009-03-26)
TfdT is a LysR-type transcriptional regulator that activates the transcription of the chlorocatechol degradative gene operon tfdCDEF of the chlorobenzoate-degrading bacterium Burkholderia sp. NK8. To identify the amino acids involved in the effector recognition by TfdT, a polymerase-chain-reaction-based random mutagenesis
Satoshi Nishino et al.
The journal of physical chemistry. A, 111(30), 7041-7047 (2007-07-03)
Intramolecular hydrogen atom tunneling in 2-chlorobenzoic acid has been investigated by low-temperature matrix-isolation infrared spectroscopy with the aid of density functional theory calculation. Infrared spectra of two relatively stable syn isomers, SC and ST, were observed in argon and xenon
C A Blackledge et al.
Journal of pharmaceutical and biomedical analysis, 22(6), 1023-1028 (2000-06-17)
The metabolic fate of [14C]-labelled 2 and 4-chlorobenzoic acids (2- and 4-CBA) has been determined in the rat following intraperitoneal (i.p.) administration at 100 mg/kg to male rats. The major route of elimination for both 2-and 4-CBA was urine with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service