Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506567

Supelco

EPA 8270/Appendix IX Semivolatile Calibration Mix

1000 μg/mL each component in dichloromethane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502

grade

analytical standard

Agency

EPA 8270

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Acetophenone
    4-Aminobiphenyl
    Aniline
    Benzyl alcohol
    m-Cresol
    2,6-Dichlorophenol
    p-Dimethylaminoazobenzene
    7,12-Dimethylbenz[a]anthracene
    1,3-Dinitrobenzene
    Dinoseb
    Diphenylamine
    Ethyl methanesulfonate
    N-(2-Fluorenyl)acetamide
    Hexachloropropene
    Isosafrol
    Methapyrilene HCl
    3-Methylcholanthrene
    Methyl methanesulfonate
    1-Naphthylamine
    2-Naphthylamine
    4-Nitroquinoline N-oxide
    N-Nitrosodibutylamine
    N-Nitrosodiethylamine
    N-Nitrosomethylethylamine
    Nitrosomorpholine
    1-Nitrosopiperidine
    1-Nitrosopyrrolidine
    2-Methyl-5-nitroaniline
    Pentachlorobenzene
    Pentachloroethane
    Pentachloronitrobenzene
    Phenacetin
    2-Picoline
    Pyridine trifluoroacetate
    Safrole
    1,2,4,5-Tetrachlorobenzene
    2,3,4,6-Tetrachlorophenol
    o-Toluidine
    1,3,5-Trinitrobenzene
    See All (39)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service