Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

505420

ASI UHMW PE pump seal

for use with M-45, 501, 510, 590, 600E, 6000

Synonym(s):

ASI HPLC Pump Part for Waters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

manufacturer/tradename

Waters 26613

packaging

1 ea of

compatibility

for use with M-45, 501, 510, 590, 600E, 6000

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service