Key Documents
503665
Regulator Wall Bracket
Single station, CGA 580
About This Item
UNSPSC Code:
41112500
feature
Single station
fitting
CGA 580
General description
Wall-mounting a gas cylinder regulator eliminates the need to handle it during cylinder change-out, thereby reducing the chance of kinking the tubing (kinked tubing next to a fitting may result in a leak). The bracket is fabricated from grade 304 stainless steel for long life, and can be used for 2-stage or 1-stage gas cylinder regulators (order separately). An integral 30 in. flexible stainless steel pigtail allows connection to a cylinder without the risk of a leak caused from kinked tubing. The CGA connector is a hand-tight design, eliminating the need for a wrench during cylinder change-out. Choose:
  • CGA 580 for helium, nitrogen, and argon
  • CGA 350 for hydrogen, ethane, and 5% methane in argon
  • CGA 590 for compressed air
The two station bracket has 2 pigtails that join before the regulator fitting, and allows 2 gas cylinders to feed the system, extending the time between cylinder change-out.
Choose from one of the most recent versions:
Certificates of Analysis (COA)