Key Documents
503363
High Purity Regulators without Purge Valve
single-stage
, CGA 350, 1/4 in. male NPT endfitting
About This Item
UNSPSC Code:
41112500
Pricing and availability is not currently available.
fitting
1/4 in. male NPT endfitting
CGA 350
Application
H2, CH2, Ar/CH4
Features and Benefits
Pressure Regulators:
High Purity and Ultra-High Purity Pressure Regulators
  • Specially cleaned to eliminate halocarbon and hydrocarbon residues
  • Filters in fitting and regulator body protect regulator from particles and pressure surges
  • Easy-to-read 2½ in. (6.3cm) gauges. Gauges calibrated in psi on regulators with CGA fittings, in bar on reguloators with DIN fittings.
  • Leak tested, ready to use.
  • High purity regulators leak tested to 2 × 10-5 cc/s (helium).
  • Ultra-high purity regulators leak tested to 2 × 10-8 cc/s (helium)
Choose from one of the most recent versions:
Certificates of Analysis (COA)