Skip to Content
MilliporeSigma
All Photos(1)

Documents

501654

Supelco

Carbon Monoxide, Carbon Dioxide, Hydrogen and Oxygen

0.5 % (w/w) each component in nitrogen, cylinder of 48 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

form

gas

packaging

cylinder of 48 L

manufacturer/tradename

Scott Gas T217-48

availability

not available in EU

concentration

0.5 % (w/w) each component in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Carbon dioxide (99.8%)
    Carbon monoxide
    Hydrogen
    Oxygen

regulator

Pictograms

Gas cylinderHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Press. Gas Compr. Gas - Repr. 1A

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service