Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49142

Supelco

2,4-D methyl ester

PESTANAL®, analytical standard

Synonym(s):

Methyl (2,4-dichlorophenoxy)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8Cl2O3
CAS Number:
Molecular Weight:
235.06
Beilstein:
2053392
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

COC(=O)COc1ccc(Cl)cc1Cl

InChI

1S/C9H8Cl2O3/c1-13-9(12)5-14-8-3-2-6(10)4-7(8)11/h2-4H,5H2,1H3

InChI key

HWIGZMADSFQMOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W A Said et al.
Applied and environmental microbiology, 57(5), 1498-1503 (1991-05-01)
Biodegradation inhibition of a benchmark chemical, 2,4-dichloro-phenoxyacetic acid methyl ester (2,4-DME), was used to quantify the inhibitory effects of heavy metals on aerobic microbial degradation rates of organic chemicals. This procedure used lake sediments and aufwuchs (floating mats) collected in
Devon J Sutherland et al.
Journal of agricultural and food chemistry, 51(1), 14-20 (2002-12-28)
The transport and fate of pesticides applied to ornamental plant nursery crops are not well documented. Methodology for analysis of soil and water runoff samples concomitantly containing the herbicides simazine (1-chloro-4,6-bis(ethylamino)-s-triazine) and 2,4-D ((2,4-dichlorophenoxy)acetic acid) was developed in this research
Qunzhou Zhang et al.
Journal of immunology (Baltimore, Md. : 1950), 183(12), 7787-7798 (2009-11-20)
Aside from the well-established self-renewal and multipotent differentiation properties, mesenchymal stem cells exhibit both immunomodulatory and anti-inflammatory roles in several experimental autoimmune and inflammatory diseases. In this study, we isolated a new population of stem cells from human gingiva, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service