Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49089

Supelco

Simazine

PESTANAL®, analytical standard

Synonym(s):

2,4-Bis(ethylamino)-6-chloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12ClN5
CAS Number:
Molecular Weight:
201.66
Beilstein:
10895
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

CCNc1nc(Cl)nc(NCC)n1

InChI

1S/C7H12ClN5/c1-3-9-6-11-5(8)12-7(13-6)10-4-2/h3-4H2,1-2H3,(H2,9,10,11,12,13)

InChI key

ODCWYMIRDDJXKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veria Addorisio et al.
Journal of hazardous materials, 196, 242-247 (2011-10-11)
Two mesoporous metal oxides, Al(2)O(3) and Fe(2)O(3), were evaluated as regards their ability to remove simazine, a highly persistent herbicide of s-triazines, using a batch equilibrium method. The effect of several experimental parameters such as pH, contact time, initial concentration
Pierre R Band et al.
The Prostate, 71(2), 168-183 (2010-08-28)
Several epidemiologic studies have reported an increased risk of prostate cancer among farmers. Our aim was to assess the risk of developing prostate cancer in relation to exposure to specific active compounds in pesticides. A case-control approach was used with
Daniel Eikel et al.
Rapid communications in mass spectrometry : RCM, 25(16), 2345-2354 (2011-07-19)
An automated surface-sampling technique called liquid extraction surface analysis (LESA), coupled with infusion nano-electrospray high-resolution mass spectrometry and tandem mass spectrometry (MS/MS), is described and applied to the qualitative determination of surface chemical residues resulting from the artificial spraying of
V Morgante et al.
Journal of environmental management, 95 Suppl, S300-S305 (2011-08-02)
Simazine is an s-triazine herbicide world widely used for the control of broadleaf weeds. The influence of leaching and microorganisms on simazine attenuation in an agricultural soil long-term treated with this herbicide was studied. To elucidate the leaching potential of
J Velisek et al.
Ecotoxicology and environmental safety, 76(2), 79-86 (2011-11-01)
The effects of a 90 day simazine exposure at concentrations of 0.06 (reported concentration in Czech rivers), 1, 2, and 4 μg L⁻¹ were assessed in one-year-old common carp (Cyprinus carpio L.). Its influence on biometric parameters, hematology, blood biochemistry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service