Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48825

Supelco

PCB Kit 3

200 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

EPA 505,508,8082,8270
MISA (Canada) Group 27

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

pkg of 1 kit

concentration

200 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

General description

Polychlorinated biphenyls are highly stable chemical products used in industries as fluids, diluents, hydraulic fluids, and dielectric fluids for capacitors and transformers.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Kit Components Only

Product No.
Description

  • Aroclor 1232 solution, certified reference material, 200 μg/mL in methanol 1 mL

  • Aroclor 1248 solution, certified reference material, 200 μg/mL in methanol 1 mL

Kit Components Also Available Separately

Product No.
Description
SDS

  • 48701Aroclor 1016 solution, certified reference material, 200 μg/mL in methanol 1 mLSDS

  • 48705Aroclor 1221 solution, certified reference material, 200 μg/mL in methanol 1 mLSDS

  • 48706Aroclor 1242 solution, certified reference material, 200 μg/mL in methanol 1 mLSDS

  • 48707Aroclor 1254 solution, certified reference material, 200 μg/mL in methanol 1 mLSDS

  • 48704Aroclor 1260 solution, certified reference material, 200 μg/mL in methanol 1 mLSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polychlorinated biphenyls: Congener-specific analysis of a commercial mixture and a human milk extract.
Safe S.
Journal of Agricultural and Food Chemistry, 33(1), 24-29 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service