Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48799-U

Supelco

EPA VOC Mix 6

2000 μg/mL each component in methanol, analytical standard

Synonym(s):

Volatile Organic Compounds Mix 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502

grade

analytical standard

Agency

EPA 601,8010,8240,8260,502.1,502.2,524.1,524.2,8021 (1987),8021A (1990)
MISA (Canada) Group 16
suitable for EPA 8260

CofA

certificate of analysis is enclosed in each package.

packaging

ampule of 1.5 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Bromomethane
    Chloroethane
    Chloromethane
    Dichlorodifluoromethane
    Trichlorofluoromethane
    Vinyl chloride

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service