Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48724-U

Supelco

Decafluorotriphenylphosphine solution

certified reference material, 25 mg/mL in dichloromethane

Synonym(s):

2,3,4,5,6,2′,3′,4′,5′,6′-Decafluorotriphenylphosphine, Bis(pentafluorophenyl)phenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 625,8240,8270

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type tuning solution

packaging

ampule of 1 mL

concentration

25 mg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C18H5F10P/c19-7-9(21)13(25)17(14(26)10(7)22)29(6-4-2-1-3-5-6)18-15(27)11(23)8(20)12(24)16(18)28/h1-5H

InChI key

OYNXPGGNQMSMTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Donnelly et al.
Journal - Association of Official Analytical Chemists, 71(2), 434-439 (1988-03-01)
Mass and intensity calibration of gas chromatograph/mass spectrometer (GC/MS) responses is an important quality assurance issue for chemical analysis. Ion abundance calibration with decafluorotriphenylphosphine (DFTPP) was applied in 1975 to standardize quadrupole spectra to resemble the ion abundances that were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service