Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48587

Supelco

Aroclor 1221

ampule of 50 mg, Neat, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Havel et al.
FEMS microbiology letters, 108(2), 211-217 (1993-04-01)
The hybrid Pseudomonas cepacia strain JHR22 was tested for its ability to degrade Aroclor 1221 in soil. The influence of supplements--mineral salts and trace elements--on the degradation was investigated. Disappearance of Aroclor 1221 congeners, occurrence of metabolites, and release of
Biodegradation of Aroclor 1221 type PCBs in sewage wastewater.
D Liu
Bulletin of environmental contamination and toxicology, 27(5), 695-703 (1981-11-01)
F Fava
Chemosphere, 32(8), 1477-1483 (1996-04-01)
A bacterial co-culture, ECO3, constituted by a polychlorobiphenyl degrading bacterium, Pseudomonas sp. strain CPE1, and two chlorobenzoic acid degrading bacteria, could grow on Aroclor 1221 (75 mg/L) as the sole carbon source without accumulating chlorinated aromatic metabolites in the medium;
F Fava et al.
Applied microbiology and biotechnology, 45(4), 562-568 (1996-05-01)
Cells of an aerobic three-membered bacterial co-culture, designated as ECO3, capable of cometabolizing and aerobically dechlorinating low-chlorinated biphenyls in the presence of biphenyl, were immobilized on Manville silica beads, on frosted-glass beads and on polyurethane foam cubes in packed-bed bioreactors
Selim Kutlu et al.
Cell biochemistry and function, 25(2), 167-172 (2005-09-24)
Polychlorinated biphenyls (PCBs) are persistent environmental pollutants. This study compared effects of two PCB mixtures, Aroclors 1221 (A1221) and 1254 (A1254) on serum levels of aspartate aminotransferase (AST), alanine aminotransferase (ALT), alkaline phosphatase (ALP), urea, creatinine and uric acid in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service