Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48557

Supelco

Bis(2-ethylhexyl) phthalate

analytical standard

Synonym(s):

DEHP, Phthalic acid bis(2-ethylhexyl ester), ‘Dioctyl’ phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Beilstein:
1890696
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

386 °C (lit.)

density

0.985 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

392.0 °F - Pensky-Martens closed cup

Flash Point(C)

200 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N B Hopf et al.
Toxicology letters, 224(1), 47-53 (2013-10-22)
Phthalates are suspected to be endocrine disruptors. Di(2-ethylhexyl) phthalate (DEHP) is assumed to have low dermal absorption; however, previous in vitro skin permeation studies have shown large permeation differences. Our aims were to determine DEHP permeation parameters and assess extent
Na Rae Song et al.
Environment international, 54, 65-73 (2013-02-19)
Di(2-ethylhexyl) phthalate (DEHP) is one of the common phthalate plasticizers used primarily in soft polyvinyl chloride, which is a plastic polymer that makes up the total weight of goods from 1% up to 40% in many consumer products. The aims
Ina Olmer Specht et al.
Environment international, 66, 146-156 (2014-03-04)
Phthalates which are widely used, are ubiquitous in the environment and in some human tissues. It is generally accepted that phthalates exert their toxic action by inhibiting Leydig cell synthesis of testosterone, but in vitro studies have also shown anti-androgenic
Anne-Marie Saillenfait et al.
Journal of applied toxicology : JAT, 33(9), 1027-1035 (2013-06-12)
In utero exposure to the phthalate ester plasticizer di-n-hexyl phthalate (DnHP) is known to affect the development of the male reproductive system and induce alterations in androgen-dependent tissues of male rat offspring. Male reproductive malformations produced by several phthalates have
Xi-Feng Zhang et al.
Environmental and molecular mutagenesis, 54(5), 354-361 (2013-04-30)
Diethylhexyl phthalate (DEHP) is an estrogen-like compound widely used as a commercial plasticizer and present in medical devices, tubing, food containers and packaging. It is considered an endocrine disruptor and studies on experimental animals showed that exposure to DEHP can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service