Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48518

Supelco

2,4,6-Trichlorophenol

analytical standard, ampule of 5000 mg

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OH
CAS Number:
Molecular Weight:
197.45
Beilstein:
776729
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

vapor pressure

1 mmHg ( 76.5 °C)

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

246 °C (lit.)

mp

64-66 °C (lit.)

application(s)

environmental

SMILES string

Oc1c(Cl)cc(Cl)cc1Cl

InChI

1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H

InChI key

LINPIYWFGCPVIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick W Causey et al.
Journal of medicinal chemistry, 51(9), 2833-2844 (2008-04-17)
A series of mono and diaryl rhenium(I)-carborane derivatives were prepared using microwave heating and screened for their affinity for two isoforms of the estrogen receptor (ER). The rhenacarborane derivative [(RR'C 2B9H9)Re(CO)3](-) (R = p-PhOH, R' = H), which was generated
Giorgio Ottaviani et al.
Journal of medicinal chemistry, 50(4), 742-748 (2007-02-16)
The development of in silico and in vitro tools to estimate or predict the passive human skin permeation and distribution of new chemical entities, useful in dermal drug delivery, in absorption studies of toxic compounds, and in the cosmetics industry
Bram Baert et al.
Bioorganic & medicinal chemistry, 15(22), 6943-6955 (2007-09-11)
A set of 116 structurally very diverse compounds, mainly drugs, was characterized by 1630 molecular descriptors. The biological property modelled in this study was the transdermal permeability coefficient logK(p). The main objective was to find a limited set of suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service