Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48358

Supelco

Acetone solution

certified reference material, 2000 μg/mL in methanol: water (9:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol: water (9:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone is a carbonyl-group bearing chemical which is categorized under the ketone family of compounds.

Application

Acetone has been used as a reference standard for the determination of the analyte in human breath, cattle breath and human blood by solid-phase extraction combined with gas chromatography–mass spectrometry (GC-MS). It may be used as a solvent in volatile phenols determination by dispersive liquid–liquid microextraction (DLLME) combined with GC-MS.(5)
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review of volatile organic compound source apportionment by chemical mass balance
Watson JG, et al.
Atmospheric Environment, 35(9), 1567 -11584 (2001)
Analysis of volatile disease markers in blood
Miekisch W, et al.
Clinical Chemistry, 47(6), 1053 -11060 (2001)
Sampling and analysis of volatile organic compounds in bovine breath by solid-phase microextraction and gas chromatography -mass spectrometry
Spinhirne JP, et al.
Journal of Chromatography A, 1025(1), 63 -669 (2004)
Determination of acetone in human breath by gas chromatography -mass spectrometry and solid-phase microextraction with on-fiber derivatization
Deng C, et al.
Journal of Chromatography. B, Biomedical Applications, 810(2), 269 -2275 (2004)
Sinisa Bjelic et al.
Journal of molecular biology, 426(1), 256-271 (2013-10-29)
Designed retroaldolases have utilized a nucleophilic lysine to promote carbon-carbon bond cleavage of β-hydroxy-ketones via a covalent Schiff base intermediate. Previous computational designs have incorporated a water molecule to facilitate formation and breakdown of the carbinolamine intermediate to give the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service