Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48268

Supelco

ASTM® D5134 Qualitative Reference Refinery Standard Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D5134

packaging

pkg of 1 kit

concentration

(2 x 1 mL each of the three standards)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Kit Components Only

Product No.
Description

  • ASTM® D5134 Qualitative Reference Alkylate Standard, analytical standard

  • ASTM® D5134 Qualitative Reference Naphtha Standard, analytical standard

Kit Components Also Available Separately

Product No.
Description
SDS

  • 48266ASTM® D5134 Qualitative Reference Reformate Standard, analytical standardSDS

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Central nervous system,Blood

Storage Class Code

3 - Flammable liquids

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service