Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48187

Supelco

Atrazine solution

1000 μg/mL in methyl tert-butyl ether, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14ClN5
CAS Number:
Molecular Weight:
215.68
Beilstein:
612020
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

CCNc1nc(Cl)nc(NC(C)C)n1

InChI

1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)

InChI key

MXWJVTOOROXGIU-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

14.0 °F

Flash Point(C)

-10 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L P Wackett et al.
Applied microbiology and biotechnology, 58(1), 39-45 (2002-02-08)
s-Triazine ring compounds are common industrial chemicals: pesticides, resin intermediates, dyes, and explosives. The fate of these compounds in the environment is directly correlated with the ability of microbes to metabolize them. Microbes metabolize melamine and the triazine herbicides such
Karsten Nödler et al.
Marine pollution bulletin, 70(1-2), 112-118 (2013-03-14)
Despite its EU-wide ban in 2004 the triazine herbicide atrazine (ATR) and its transformation products (TPs) are frequently detected in the aqueous environment. In this study, concentrations of ATR, two of its TPs, and terbuthylazine (TERB) are presented and discussed
Keith R Solomon et al.
Critical reviews in toxicology, 38(9), 721-772 (2008-10-23)
The herbicide atrazine is widely used in agriculture for the production of corn and other crops. Because of its physical and chemical properties, atrazine is found in small concentrations in surface waters--habitats for some species. A number of reports on
M Graymore et al.
Environment international, 26(7-8), 483-495 (2001-08-04)
A portion of all herbicides applied to forests, croplands, road sides, and gardens are inevitably lost to water bodies either directly through runoff or indirectly by leaching through groundwater into ephemeral streams and lakes. Once in the aquatic environment, herbicides
T Komang Ralebits et al.
Biodegradation, 13(1), 11-19 (2002-09-12)
The potential toxicity of the s-triazine herbicide atrazine motivates continuous bioremediation-directed research. Several indigenous soil atrazine-catabolizing microbial associations and monocultures have been enriched/isolated from compromised sites. Of these, Pseudomonas sp. strain ADP has become a reference strain and has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service