Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47841

Supelco

D-Sorbitol

analytical standard

Synonym(s):

D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721899
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E420

grade

analytical standard

vapor density

<1 (vs air)

vapor pressure

<0.1 mmHg ( 25 °C)

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

98-100 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Other Notes

Ion Mobility: TWCCSN2 value of 139.6 Å2 [M+Na]+ and 132.3 Å2 [M-H]-

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 47841 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective determination of D-sorbitol and D-mannitol in foodstuffs by ion chromatography with polarized photometric detection
Yamamoto A, et al.
Journal of Chromatography A, 804(1-2), 305-309 (1998)
J M van Griensven et al.
Clinical pharmacology and therapeutics, 58(6), 631-640 (1995-12-01)
To examine the effect of diabetes mellitus on the pharmacokinetics of tolrestat and to investigate its effect on red blood cell sorbitol levels according to a new pharmacodynamic model for this class of drugs. Single and multiple doses of tolrestat
Youngkook Kwon et al.
ChemSusChem, 6(3), 455-462 (2013-01-25)
This Full Paper addresses the electrocatalytic hydrogenation of glucose to sorbitol or 2-deoxysorbitol on solid metal electrodes in neutral media. Combining voltammetry and online product analysis with high-performance liquid chromatography (HPLC), provides both qualitative and quantitative information regarding the reaction
Anna Boccaccio et al.
Cellular and molecular life sciences : CMLS, 71(21), 4275-4283 (2014-04-29)
Two-pore channel proteins (TPC) encode intracellular ion channels in both animals and plants. In mammalian cells, the two isoforms (TPC1 and TPC2) localize to the endo-lysosomal compartment, whereas the plant TPC1 protein is targeted to the membrane surrounding the large
Arie Gruzman et al.
Journal of medicinal chemistry, 51(24), 8096-8108 (2008-12-04)
Type 2 diabetes mellitus has reached epidemic proportions; therefore, the search for novel antihyperglycemic drugs is intense. We have discovered that D-xylose increases the rate of glucose transport in a non-insulin-dependent manner in rat and human myotubes in vitro. Due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service