Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47840

Supelco

Saccharin Calcium Salt

analytical standard

Synonym(s):

Saccharin hemicalcium salt, 2,3-Dihydro-3-oxobenzisosulfonazole, o-Sulfobenzimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8CaN2O6S2 · 3.5H2O
CAS Number:
Molecular Weight:
467.48
UNSPSC Code:
12164500
E Number:
E954

grade

analytical standard

Quality Level

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

room temp

InChI

1S/2C7H5NO3S.Ca.H2O/c2*9-7-5-3-1-2-4-6(5)12(10,11)8-7;;/h2*1-4H,(H,8,9,10,11);;1H2/q;;+2;/p-2

InChI key

XOCXGVIOTKFUSU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service