Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47749-U

Supelco

2-Amino-4,6-dinitrotoluene solution

certified reference material, 1000 μg/mL in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3O4
CAS Number:
Molecular Weight:
197.15
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 8330

feature

standard type calibration

concentration

1000 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C7H7N3O4/c1-4-6(8)2-5(9(11)12)3-7(4)10(13)14/h2-3H,8H2,1H3

InChI key

IEEJAAUSLQCGJH-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G S Nyanhongo et al.
Journal of hazardous materials, 165(1-3), 285-290 (2008-11-18)
Pseudomonas putida GG04 and Bacillus SF have been successfully incorporated into an explosive formulation to enhance biotransformation of TNT residues and/or explosives which fail to detonate due to technical faults. The incorporation of the microorganisms into the explosive did not
Alethea T Bowen et al.
Chemosphere, 63(1), 58-63 (2005-12-06)
Tubifex tubifex metabolizes 2,4,6-trinitrotoluene (TNT) to 2-amino-4,6-dinitrotoluene (2ADNT) and 4-amino-2,6-dinitrotoluene (4ADNT). Elimination rates of metabolically-generated ADNTs are low compared to ADNTs absorbed directly from water, suggesting that metabolically-generated ADNTs may be bound or sequestered within tissue and therefore less available
L R Krumholz et al.
Journal of industrial microbiology & biotechnology, 18(2-3), 161-169 (1997-02-01)
The transport and fate of pollutants is often governed by both their tendency to sorb as well as their susceptibility to biodegradation. We have evaluated these parameters for 2,4,6-trinitrotoluene (TNT) and several biodegradation products. Slurries of aquifer sediment and groundwater
Craig A McFarland et al.
Environmental pollution (Barking, Essex : 1987), 159(2), 466-473 (2010-11-12)
The compound 2-amino-4,6-dinitrotoluene (2A-DNT) was evaluated under laboratory conditions in the Western fence lizard (Sceloporus occidentalis) to assess the potential for reptile toxicity. Oral LD(50) values were 1406 and 1867 mg/kg for male and female lizards, respectively. Based on responses
Michael J Quinn et al.
Ecotoxicology (London, England), 19(5), 945-952 (2010-03-10)
2-Amino-4,6-dinitrotoluene (2A-DNT) is a metabolite of the explosive 2,4,6-trinitrotoluene (TNT) which is present in the soil at numerous U.S. Army installations as the result of TNT manufacture or training activities. Although many avian species are known to inhabit areas where

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service