Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47323

Supelco

Propylbenzene

analytical standard

Synonym(s):

1-Phenylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CH3
CAS Number:
Molecular Weight:
120.19
Beilstein:
1903006
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.14 (vs air)

vapor pressure

2 mmHg ( 20 °C)

autoignition temp.

842 °F

expl. lim.

6 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.491 (lit.)

bp

159 °C (lit.)

mp

−99 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

CCCc1ccccc1

InChI

1S/C9H12/c1-2-6-9-7-4-3-5-8-9/h3-5,7-8H,2,6H2,1H3

InChI key

ODLMAHJVESYWTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Cruz et al.
British journal of pharmacology, 131(7), 1303-1308 (2000-11-23)
1. We have previously shown that toluene dose-dependently inhibits recombinant N-methyl-D-aspartate (NMDA) receptors at micromolar concentrations. This inhibition was rapid, almost complete and reversible. The NR1/2B combination was the most sensitive receptor subtype tested with an IC(50) value for toluene
R Andrew Shalliker et al.
Journal of chromatography. A, 1216(5), 787-793 (2008-12-20)
The effect of solvent viscosity mismatch on elution performance in reversed-phase HPLC was studied using moment analysis. Two conditions were tested: (1) the mobile phase viscosity was less than the injection plug viscosity, and (2) the mobile phase viscosity was
Sara Eriksson et al.
Biodegradation, 16(3), 253-263 (2005-05-04)
The metabolism of monoaromatic hydrocarbons by an iron-reducing bacterial enrichment culture originating from diesel-contaminated groundwater was examined using d7-propylbenzene as a model hydrocarbon. Sequence analysis of the 16S rDNA gene showed that the dominant part (10 of 10 clones) of
Matthew Giardina et al.
Analytical chemistry, 75(7), 1604-1614 (2003-04-23)
With increasing interest in the detection of disease-related volatile organic compounds (VOCs) found in human breath, breath analysis could prove to be a very useful diagnostic tool, especially for the early detection of lung cancer. Solid-phase microextraction (SPME) is a
Y C Shan et al.
Se pu = Chinese journal of chromatography, 19(3), 256-259 (2003-01-25)
A new method for predicting the retention time of solutes under linear gradient elution has been established. In this method, the migration distance of solutes in column and the mobile phase gradient were modified simultaneously according to the distribution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service