Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46916-U

Supelco

Zearalenone solution

50 μg/mL in acetonitrile, analytical standard

Synonym(s):

ZON

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O5
CAS Number:
Molecular Weight:
318.36
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

packaging

ampule of 1 mL

concentration

50 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

OC1=C(C(O[C@@H](C)CCC2)=O)C(/C=C/CCCC2=O)=CC(O)=C1

InChI

1S/C18H22O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,20-21H,2,4-6,8-9H2,1H3/b7-3+/t12-/m0/s1

InChI key

MBMQEIFVQACCCH-QBODLPLBSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

General description

Zearalenone is a mycotoxin, produced by Fusarium species and often contaminates grains and stored feeds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Zearalenone was used as standard in microwave-assisted extraction method for determination of zearalenone in wheat and corn by liquid chromatography-mass spectrometry with an atmospheric pressure chemical ionization interface. It was used as analytical standard in determination of zearalenone content of maize, wheat, barley, swine feed and poultry feed samples by immunoaffinity column clean-up followed by liquid chromatography. It may be used as internal standard in simultaneous determination of the type A- and B-trichothecenes as well as zearalenone in maize by high performance liquid chromatography-tandem mass spectrometry.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Fazekas et al.
Journal of AOAC International, 84(5), 1453-1459 (2001-10-17)
The zearalenone content of maize, wheat, barley, swine feed, and poultry feed samples was determined by immunoaffinity column cleanup followed by liquid chromatography (IAC-LC). Samples were extracted in methanol-water (8 + 2, v/v) solution. The filtered extract was diluted with
Lea Pallaroni et al.
Analytical and bioanalytical chemistry, 374(1), 161-166 (2002-09-11)
A microwave-assisted extraction (MAE) method has been developed for determination of zearalenone in wheat and corn by LC-MS with an atmospheric pressure chemical ionization interface (APCI). Matrix effects were minimized by use of matrix-matched standard curves for quantification of the
Franz Berthiller et al.
Journal of chromatography. A, 1062(2), 209-216 (2005-02-01)
A novel method for the simultaneous determination of the Fusarium mycotoxins nivalenol, deoxynivalenol, fusarenon-X, 3-acetyl-deoxynivalenol, the sum of 3-acetyl-deoxynivalenol and 15-acetyl-deoxynivalenol, diacetoxy-scirpenol, HT-2 toxin, T-2 toxin and zearalenone in maize has been developed using gradient RP-LC with atmospheric pressure chemical
Cristina Juan et al.
Food chemistry, 134(4), 2389-2397 (2013-02-28)
Although analytical methods have been already reported for legislated mycotoxins as trichothecenes and zearalenone (ZON) separately, we describe the optimization of a simple and rapid multimycotoxin method for the determination of a total of 12 mycotoxins simultaneously, nine trichothecenes (NIV
Fei Zhao et al.
Toxicological sciences : an official journal of the Society of Toxicology, 132(2), 431-442 (2013-01-08)
Zearalenone (ZEA) is a mycotoxin commonly found in contaminated livestock feed and human food with levels in the range of ppb and low ppm. It was hypothesized that ZEA, an endocrine disruptor, could affect puberty and early pregnancy. To test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service