Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44585-U

Supelco

n-Paraffins Mix

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Agency

ASTM® D6729,D6730,D6733

packaging

pkg of 0.1 mL

concentration

(typical values shown)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

General description

This is an eleven component mix of known paraffins. It is intended for use in identifying paraffins in complex hydrocarbon mixtures.

  • Formulation is weight percent, and will vary lot to lot.
  • A detailed data sheet listing components by weight percent, mole percent, and liquid volume percent, retention times and other formulaiton is provided with the product.
  • Supplied in a crimp-top vial with hole cap and septa.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Pentane 9.38 wt. %
    Hexane 9.508 wt. %
    Heptane 9.776 wt. %
    Octane 9.544 wt. %
    Nonane 9.042 wt. %
    Decane 9.245 wt. %
    Undecane 9.313 wt. %
    Dodecane 9.404 wt. %
    Tridecane 8.907 wt. %
    Tetradecane 8.792 wt. %
    Pentadecane 7.087 wt. %
    See All (11)

analytical column

standard

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-57.0 °F

Flash Point(C)

-49.45 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabiane M Nachtigall et al.
Foods (Basel, Switzerland), 8(12) (2019-11-24)
The objective of this study was to investigate the reduction and partial substitution effects of sodium chloride (NaCl) by potassium chloride (KCl) and calcium chloride (CaCl2) on lipolysis and lipid oxidation in salted meat aiming at reducing sodium content. To
G Kaban et al.
Poultry science, 99(4), 2293-2299 (2020-04-04)
Salted-dried goose is a traditional Turkish product with specific flavor that is produced by dry salting, post-salting, and subsequently air-drying of the goose carcass. In this study, the leg and breast parts of salted-dried goose carcasses were analyzed in terms
Noemi Echegaray et al.
Foods (Basel, Switzerland), 9(6) (2020-06-11)
The aim of this study was to evaluate the influence of the diet (chestnut vs. commercial feed) and cooking techniques (roasting, grilling, frying and microwaving) on the quality of the Biceps femoris muscle of the Celta pig breed. Chemical composition
Ahmet Akköse et al.
Asian-Australasian journal of animal sciences, 30(8), 1168-1174 (2016-12-23)
The aim of the study was to evaluate the effects of different nitrate levels (150, 300, 450, and 600 ppm KNO Pastırma samples were produced under the controlled condition and analyses of volatile compounds, and thiobarbituric acid reactive substances (TBARS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service