Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442541

Supelco

DEET

PESTANAL®, analytical standard

Synonym(s):

N,N-Diethyl-m-toluamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
Beilstein:
2046711
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

CCN(CC)C(=O)c1cccc(C)c1

InChI

1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3

InChI key

MMOXZBCLCQITDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DEET can find applications as a chemical repellent in order to control a wide range of insects like bed bugs, mosquitos and fruit flies. Its mode of action basically involves the interference with the odorant receptors leading to avoidance kind of behaviour.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicity, repellency and flushing out in Triatoma infestans (Hemiptera: Reduviidae) exposed to the repellents DEET and IR3535
Reynoso.N.MM, et al.
PeerJ, 5 (2017)
Molecular Basis of N, N-Diethyl-3-Methylbenzamide (DEET) in Repelling the Common Bed Bug, Cimex lectularius
Liu F, et al.
Frontiers in Physiology, 8 (2017)
P J Robbins et al.
Journal of toxicology and environmental health, 18(4), 503-525 (1986-01-01)
A review of the biodistribution and toxicity of the insect repellent N,N-diethyl-m-toluamide (DEET) is presented. Workers using repellent containing this compound may be exposed to greater than 442 g in 6 mo. In human studies, variable penetration into the skin
Pinky Kain et al.
Nature, 502(7472), 507-512 (2013-10-04)
There are major impediments to finding improved DEET alternatives because the receptors causing olfactory repellency are unknown, and new chemicals require exorbitant costs to determine safety for human use. Here we identify DEET-sensitive neurons in a pit-like structure in the
Daniel L Sudakin et al.
Journal of toxicology. Clinical toxicology, 41(6), 831-839 (2003-12-18)
The emergence of West Nile Virus (WNV) in North America has resulted in increased public awareness and utilization of insect repellents containing N,N-diethyl-m-toluamide (DEET) in the prevention of mosquito-borne disease. Regulatory reassessments in North American countries have recently been completed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service