Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442530

Supelco

Cyanazine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13ClN6
CAS Number:
Molecular Weight:
240.69
Beilstein:
615509
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

CCNc1nc(Cl)nc(NC(C)(C)C#N)n1

InChI

1S/C9H13ClN6/c1-4-12-7-13-6(10)14-8(15-7)16-9(2,3)5-11/h4H2,1-3H3,(H2,12,13,14,15,16)

InChI key

MZZBPDKVEFVLFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Borja Muñoz-Solano et al.
Toxins, 12(6) (2020-06-11)
Mycotoxins are toxic compounds for humans and animals that are produced by fungi. Mycotoxin contamination in feed is a global safety concern and effective control of these compounds in this matrix is needed. This study proposes a simple, cost-effective analytical
Gaurav Saini et al.
Journal of chromatography. A, 1216(16), 3587-3593 (2009-01-03)
In spite of advances in solid-phase extraction (SPE) technology there are certain disadvantages to current SPE silica-based, column packings. The pH range over which extraction can occur is limited and each column is generally only used once. New diamond-based reversed
Shannon M Lynch et al.
Environmental health perspectives, 114(8), 1248-1252 (2006-08-03)
Cyanazine is a common pesticide used frequently in the United States during the 1980s and 1990s. Animal and human studies have suggested that triazines may be carcinogenic, but results have been mixed. We evaluated cancer incidence in cyanazine-exposed pesticide applicators
Ying Jin-Clark et al.
Environmental toxicology and chemistry, 21(3), 598-603 (2002-03-07)
Toxicities of two triazine herbicides (atrazine and cyanazine) and an organophosphate insecticide (chlorpyrifos) were evaluated individually and with each herbicide in binary combination with chlorpyrifos using fourth-instar larvae of the aquatic midge, Chironomus tentans. Chlorpyrifos at 0.25 microg/L resulted in
Cynthia V Rider et al.
Environmental toxicology and chemistry, 29(9), 2064-2071 (2010-09-08)
Reproductive abnormalities in alligators exposed to contaminants in Lake Apopka, Florida, USA represent a clear example of endocrine disruption in wildlife. Several of these contaminants that are not able to bind to mammalian estrogen receptors (such as atrazine and cyanazine)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service