Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442436

Supelco

Acetone-2,4-DNPH

analytical standard

Synonym(s):

Acetone-2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20
Beilstein:
960878
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

126-128 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

C\C(C)=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3

InChI key

YGIXYAIGWMAGIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone-2,4-DNPH can be obtained as an oxidation product of the reaction of alpha-pinene and hydroxyl radicals, which is determined using gas chromatography (GC) and high performance liquid chromatography (HPLC) coupled with mass spectrometry (MS).[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of the oxidation products of the reaction between ?-pinene and hydroxyl radicals by gas and high-performance liquid chromatography with mass spectrometric detection
Bergh DVV, et al.
Journal of Chromatography A, 896, 135-148 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service