Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442432

Supelco

Acenaphthene-d10

analytical standard

Synonym(s):

1,8-·Ethylenenaphthalene-D10, Deuterated acenaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D10
CAS Number:
Molecular Weight:
164.27
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

277-279 °C (lit.)

mp

95-97 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+10

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c2c3c(c([2H])c([2H])c([2H])c3c1[2H])C([2H])([2H])C2([2H])[2H]

InChI

1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2/i1D,2D,3D,4D,5D,6D,7D2,8D2

InChI key

CWRYPZZKDGJXCA-WHUVPORUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acenaphthene-d10 has been used as internal standard in the determination of pesticides in rice samples using a suitable multi-residue method such as quick, easy, cheap, effective, rugged and safe (QuEChERS) sample preparation method combined with gas chromatography coupled with mass spectrometry analysis. It can also find applications as a polycyclic aromatic hydrocarbon surrogate in the analysis of polycyclic aromatic hydrocarbons in soil, river water and sediments using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A multi-residue method for the determination of 203 pesticides in rice paddies using gas chromatography/mass spectrometry
Nguyen DT, et al.
Analytica Chimica Acta, 619, 67-74 (2008)
Levels, sources and potential risks of polycyclic aromatic hydrocarbons (PAHs) in multimedia environment along the Jinjiang River mainstream to Quanzhou Bay, China
Yang D, et al.
Marine Pollution Bulletin, 76, 298-306 (2013)
Tirumuru V Reddy et al.
Environmental toxicology and chemistry, 34(12), 2796-2802 (2015-06-20)
Endocrine-disrupting chemicals (EDCs) such as 17α-ethynylestradiol, 17β-estradiol, estrone, and para-nonylphenol have been measured in wastewater-treatment plant effluents, surface waters, sediments, and sludge and have been shown to induce liver-specific vitellogenin (vtg) messenger RNA in male fathead minnows (Pimephales promelas). The
Askin Birgul et al.
Archives of environmental contamination and toxicology, 68(1), 46-63 (2014-12-07)
Ambient air polycyclic aromatic hydrocarbon (PAH) samples were collected at traffic, residential, coastal, and semiurban sites in Bursa, Turkey, between June 2008 and June 2009. For the traffic, residential, coastal, and semiurban sites, the average gas phase total PAH (∑12PAH)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service