Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442410-U

Supelco

4-Chloroaniline

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
Beilstein:
471359
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

4.4 (vs air)

vapor pressure

0.15 mmHg ( 25 °C)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

232 °C (lit.)

mp

67-70 °C (lit.)

application(s)

environmental
pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(Cl)cc1

InChI

1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

QSNSCYSYFYORTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloroaniline is a derivative of aniline that is extensively employed as an intermediate in the production of dyes, rubbers, plastics, agrochemicals and pharmaceuticals.

Application

4-Chloroaniline may be used as an analytical reference standard for the determination of the analyte in water, synthetic food color additives, textile samples and forestry matrices by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast separation and sensitive detection of carcinogenic aromatic amines by reversed-phase ?-liquid chromatography coupled with electrochemical detection
Shelke M, et al.
Journal of Chromatography A, 1089(1-2), 52-58 (2005)
Analysis of primary aromatic amines using precolumn derivatization by HPLC fluorescence detection and online MS identification
Zhao X and Suo Y
Journal of Separation Science, 31(4), 646-658 (2008)
Analysis of aromatic amines in water samples by liquid-liquid-liquid microextraction with hollow fibers and high-performance liquid chromatography
Zhao L, et al.
Journal of Chromatography A, 963(1-2), 239-248 (2002)
HPLC/diode-array method for the determination of the pesticide diflubenzuron and its major metabolites 2,6-difluorobenzamide, 4-chlorophenylurea, and 4-chloroaniline in forestry matrixes.
Rodriguez E, et al.
Journal of Liquid Chromatography and Related Technologies, 21(12), 1857-1870 (1998)
Rapid qualitative pyrolysis GC-MS analysis of carcinogenic aromatic amines from dyed textiles
Plum A
Chromatographia, 57(1), S243-S248 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service