Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442236

Supelco

1,3,5-Trimethylbenzene

analytical standard

Synonym(s):

Mesitylene, 1,3,5-Trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
906806
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.1 (vs air)

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)

autoignition temp.

1022 °F

expl. lim.

0.88-6.1 %, 100 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.499 (lit.)

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

environmental

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth L Hommel et al.
The Analyst, 128(6), 750-755 (2003-07-18)
The air-liquid interface and the liquid-phase of benzene, toluene, 1,3-dimethylbenzene, and 1,3,5-trimethylbenzene are studied using broad bandwidth sum frequency generation spectroscopy, Raman and infrared spectroscopy. A vibrationally resonant sum frequency response is observed from these surfaces in spite of the
Eric J Werner et al.
Contrast media & molecular imaging, 4(5), 220-229 (2009-10-20)
A series of new Gd(III) hydroxypyridonate complexes featuring a mesitylene (ME)-derived ligand cap has been prepared. Relaxometric characterization reveals that the complexes tend to form large aggregates in solution with slow tumbling rates, as estimated from NMRD analysis, and unique
Zachariah M Heiden et al.
Inorganic chemistry, 50(4), 1470-1479 (2011-01-28)
The series of o-benzylphosphino-boranes, o-(R(2)B)C(6)H(4)CH(2)PtBu(2) (R = Cl 3, Ph 4, Cy 6, C(6)F(5) 7, Mes 8) and o-(BBN)C(6)H(4)CH(2)PtBu(2) (5), were synthesized from reactions of the respective chloroboranes with the lithiated benzyl-phosphine. In an analogous fashion, the α-methylbenzyl(N,N-dimethyl)amine-boranes o-(R(2)B)C(6)H(4)CH(Me)NMe(2) (R
Michael Cushion et al.
Dalton transactions (Cambridge, England : 2003), (9)(9), 1632-1635 (2009-05-08)
The new sterically encumbered facially coordinating N(3)-donor ligand cis,cis-1,3,5-tris(mesitylideneimino)cyclohexane (L1) has been synthesised. Reaction with [Cu(NCCH(3))(4)]PF(6) gives [Cu(L1)NCCH(3)]PF(6) (1), the bound acetonitrile being labile and readily replaced by CO to yield [Cu(L1)CO]PF(6) (2); both 1 and 2 have been structurally
John W Colson et al.
Science (New York, N.Y.), 332(6026), 228-231 (2011-04-09)
Covalent organic frameworks (COFs), in which molecular building blocks form robust microporous networks, are usually synthesized as insoluble and unprocessable powders. We have grown two-dimensional (2D) COF films on single-layer graphene (SLG) under operationally simple solvothermal conditions. The layered films

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service