Skip to Content
MilliporeSigma
All Photos(1)

Documents

42704

Supelco

n-Undecane

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Undecane, n-Undecane, Hendecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31
Beilstein:
1697099
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

vapor density

5.4 (vs air)

vapor pressure

<0.4 mmHg ( 20 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.417 (lit.)

bp

196 °C (lit.)

mp

−26 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCC

InChI

1S/C11H24/c1-3-5-7-9-11-10-8-6-4-2/h3-11H2,1-2H3

InChI key

RSJKGSCJYJTIGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service