Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42154

Supelco

Dodecanoic-1,2,3,4-13C4 acid

analytical standard

Synonym(s):

Lauric acid-1,2,3,4-13C4

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$214.00
10 MG
$789.00

$214.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$214.00
10 MG
$789.00

About This Item

Empirical Formula (Hill Notation):
13C4C8H24O2
CAS Number:
Molecular Weight:
204.29
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.13

$214.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99%

storage temp.

2-8°C

SMILES string

O[13C]([13CH2][13CH2][13CH2]CCCCCCCC)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i9+1,10+1,11+1,12+1

InChI key

POULHZVOKOAJMA-BMHINFNASA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service