Skip to Content
MilliporeSigma
All Photos(1)

Documents

40117-U

Supelco

Pyridine-2-Aldehyde-DNPH Standard

1 mM in acetonitrile (aldehyde, equivalent), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 3 × 2 mL

concentration

1 mM in acetonitrile (aldehyde, equivalent)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

General description

Pyridine-2-Aldehyde-DNPH Standard is used for ozone analysis. It is mainly designed as a quantitative calibration mixture where a multi-component solution is not suitable. Packaged in an amber glass ampule.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service