Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40074

Supelco

Chrysene solution

certified reference material, 1000 μg/mL in acetone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H

InChI key

WDECIBYCCFPHNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daoud Ali et al.
Toxicology letters, 204(2-3), 199-207 (2011-05-17)
Chrysene is one of the basic polycyclic aromatic hydrocarbon (PAH) which is toxic environmental pollutant and consistently exposed to sunlight. However, little information is available on its photogenotoxicity. The objective of the present study was to analyze the effects of
A I Alajtal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(1), 1-5 (2010-03-24)
Raman spectroscopy, with visible laser (514 and 633 nm) and near infrared (785 and 1064 nm) excitation, has been used to obtain high quality spectra of phenanthrene, chrysene, and tetracene. Samples with dimensions from a minimum size of 10 microm
Seiichi Uno et al.
Environmental monitoring and assessment, 165(1-4), 501-515 (2009-05-19)
Following the oil spill accident of the Solar I tanker in 2006 off the coast of Guimaras Island in the Philippines, polycyclic aromatic hydrocarbons (PAHs) and alkylated PAHs in some aquatic organisms were investigated at Luzaran in Guimaras and Taklong
Friedrich Roth et al.
The Journal of chemical physics, 137(11), 114508-114508 (2012-09-25)
We have performed electron energy-loss spectroscopy studies in order to investigate the electronic properties of chrysene molecular solids. The valence band electronic excitation spectra and the C 1s core level excitations have been measured for pristine and potassium doped chrysene.
Theodoros Tzamkiozis et al.
Inhalation toxicology, 22 Suppl 2, 59-69 (2010-10-30)
This study presents different research techniques linked together to improve our understanding of the particulate matter (PM) impacts on health. PM samples from the exhaust of different vehicles were collected by a versatile aerosol concentration enrichment system (VACES). Waterborne PM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service