Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40070-U

Supelco

Benz[a]anthracene solution

certified reference material, 1000 μg/mL in methanol

Synonym(s):

Benzo[a]anthracene solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909298
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

c1ccc2cc3c(ccc4ccccc34)cc2c1

InChI

1S/C18H12/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1-12H

InChI key

DXBHBZVCASKNBY-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luc Schuler et al.
Applied microbiology and biotechnology, 83(3), 465-475 (2009-01-28)
Sphingomonas sp. strain LH128 was isolated from a polycyclic aromatic hydrocarbon (PAH)-contaminated soil using phenanthrene as the sole source of carbon and energy. A dioxygenase complex, phnA1fA2f, encoding the alpha and beta subunit of a terminal dioxygenase responsible for the
Atika Jaruchotikamol et al.
Toxicology and applied pharmacology, 224(2), 156-162 (2007-09-11)
The effects of andrographolide, the major diterpenoid constituent of Andrographis paniculata, on the expression of cytochrome P450 superfamily 1 members, including CYP1A1, CYP1A2, and CYP1B1, as well as on aryl hydrocarbon receptor (AhR) expression in primary cultures of mouse hepatocytes
T Kido et al.
Environmental toxicology, 28(1), 21-30 (2011-03-09)
Chlorinated benz[a]anthracenes (Cl-BaA) are halogenated aromatic compounds (typified by dioxins) found in the environment at relatively high concentrations. Fischer 344 rats were intragastrically administered 0, 1, or 10 mg of Cl-BaA or its parent compound benz[a]anthracene (BaA) per kg of
Maiysha D Jones et al.
Environmental microbiology, 13(10), 2623-2632 (2011-05-14)
The bacteria responsible for the degradation of naphthalene, phenanthrene, pyrene, fluoranthene or benz[a]anthracene in a polycyclic aromatic hydrocarbon (PAH)-contaminated soil were investigated by DNA-based stable-isotope probing (SIP). Clone libraries of 16S rRNA genes were generated from the (13) C-enriched ('heavy')
Sona Marvanová et al.
Chemical research in toxicology, 21(2), 503-512 (2008-01-22)
Monomethylated benz[ a]anthracenes (MeBaAs) are an important group of methylated derivatives of polycyclic aromatic hydrocarbons (PAHs). Although the methyl substitution reportedly affects their mutagenicity and tumor-initiating activity, little is known about the impact of methylation on the effects associated with

Articles

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service