Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33318-U

Supelco

Block heater (without block), 220V

AC/DC input 230 V AC, European 2-pin plug

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,280.00

$1,280.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,280.00

About This Item

UNSPSC Code:
12352200
NACRES:
NB.77

$1,280.00


Please contact Customer Service for Availability

Request a Bulk Order

General description

This multi-purpose unit is ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures. Economical, versatile and compact. It accepts separate interchangeable modular heating blocks which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes.
Each block has a thermometer well for measuring block termperature. Heats from ambient to 150°C with two temperature adjustments, low range ambient to
100°C/high range from 75°C to 150°C.
The anodized aluminum modular heating blocks provide superior temperature stability and heat transfer.
Heating blocks are sold seperately.

Features and Benefits

  • Compact design takes up little bench space
  • Interchangeable heating blocks for maximum versatility

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hei! is the size of the aluminum heating block that fits the device is this one 3.75 in. × 2.9 in. × 2.0in? We do not need the block itself, but we need to know if the one we have in use fits in this instrument.

    1 answer
    1. The dimensions of the heating block are as follows:
      9.5 cm length (3.74")
      7.5 cm width (2.983")
      5.0 cm height (1.96")

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service