Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33175

Supelco

Silica Column Regeneration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

description

Silica Column Regeneration Solution

packaging

pkg of 200 mL

compatibility

Restoring Column Performance

storage temp.

room temp

General description

Silica Column Regeneration Solution contains a mixture of acetic acid, dimethoxypropane, and methylene chloride.
This solution effectively regenerates a silica column that has come into contact with very strongly polar solvents, such as water or alcohols. Simply flush the column with regeneration solution for 10 minutes, then reequilibrate with mobile phase for 10 minutes. Column performance usually is restored to that obtained before exposure to the polar solvent.

Application

It was used to flush and regenerate the column used for HPLC analysis of tocopherols and tocotrienols.

Analyte

Description

    Acetic acid
    Dichloromethane
    2,2-Dimethoxypropane

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple solutions to problems encountered in quantitative analysis of tocopherols and tocotrienols using silica columns.
A K Hewavitharana
Analytical biochemistry, 313(2), 342-344 (2003-02-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service