Skip to Content
MilliporeSigma
All Photos(1)

Documents

33095

Supelco

Methanolic HCl

0.5 M HCl in methanol (0.5N), for GC derivatization, ampule of 10 × 5 mL

Synonym(s):

Hydrogen chloride, Hydrochloric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
UNSPSC Code:
41116105
PubChem Substance ID:

grade

for GC derivatization

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

packaging

ampule of 10 × 5 mL

concentration

0.5 M HCl in methanol (0.5N)

storage temp.

2-8°C

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanolic HCl (hydrochloric acid in methanol) is particularly useful for preparing methyl esters of volatile (short chain) fatty acids. Fatty acids are esterified by heating them with an anhydrous alcohol (e.g., methanol) in the presence of an acidic catalyst (e.g., HCl) in a sealed vessel at a high temperature for a short time. In the reaction, a fatty acid molecule and an alcohol molecule are joined, with the release of a water molecule. The derivatives can be quickly and easily recovered, quantitatively, from the reaction medium.

Features and Benefits

  • Derivatization of fatty acids, particularly volatile (short chain) fatty acids.

  • Clean reaction (no side reaction) with volatile by-products.

  • Provides clean, fast, quantitative derivatization.

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Qiu et al.
The Journal of biological chemistry, 276(34), 31561-31566 (2001-06-09)
The existence of Delta 4 fatty acid desaturation in the biosynthesis of docosahexanoic acid (DHA) has been questioned over the years. In this report we describe the identification from Thraustochytrium sp. of two cDNAs, Fad4 and Fad5, coding for Delta
Practical Gas Chromatography: A Comprehensive Reference.
Wilder KD and Engewald W.
Science, 613-614 (2014)
Mitochondrial Function, Part B: Mitochondrial Protein Kinases, Protein Phosphatases and Mitochondrial Diseases.
William SA and Murphy AN.
Science, 88-89 (2009)
[Fluid and electrolyte movements through the gastrointestinal membranes: hydrochloric acid formation and intestinal water transport].
D P Mertz
Klinische Wochenschrift, 45(2), 57-68 (1967-01-15)
Vanessa Zambelli et al.
Anesthesia and analgesia, 120(2), 381-388 (2014-12-17)
Because pulmonary endogenous surfactant is altered during acute respiratory distress syndrome, surfactant replacement may improve clinical outcomes. However, trials of surfactant use have had mixed results. We designed this animal model of unilateral (right) lung injury to explore the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service